siRNA Details
Gene Symbol: Cldn23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027998.4 | NP_082274.1 | 1 | 967 | ||
GenBank mRNA | AK009330.1 | - | 1 | 888 | ||
AK037108.1 | - | 1 | 922 | |||
AK145246.1 | - | 1 | 971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
