siRNA Details
Gene Symbol: Tsc22d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081229.1 | NP_001074698.1 | 4 | 2340 | ||
GenBank mRNA | AK007760.1 | - | 3 | 683 | ||
AK156074.1 | - | 2 | 324 | |||
BC058221.1 | - | 3 | 536 | |||
BC117998.1 | - | 4 | 2069 | 40095084 |

More Information
Other Species Targeted

Gene Ontology Categories:
