siRNA Details
Gene Symbol: Cnot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037846.3 | NP_001032935.2 | 6 | 603 | ||
NM_001037847.2 | NP_001032936.2 | 6 | 533 | |||
NM_028082.2 | NP_082358.2 | 5 | 480 | |||
GenBank mRNA | AK011231.1 | - | 4 | 403 | ||
AK149767.1 | - | 6 | 577 | |||
AK167834.1 | - | 6 | 581 | |||
AK205222.1 | - | 2 | 135 | |||
BC043133.1 | - | 6 | 504 | |||
BC063105.1 | - | 7 | 746 | 5003026 | ||
BC065171.1 | - | 6 | 533 | 6415034 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
