siRNA Details
Gene Symbol: Cnot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037846.3 | NP_001032935.2 | 6 | 623 | ||
NM_001037847.2 | NP_001032936.2 | 6 | 553 | |||
NM_028082.2 | NP_082358.2 | 5 | 500 | |||
GenBank mRNA | AK011231.1 | - | 4 | 423 | ||
AK078121.1 | - | 6 | 613 | |||
AK149767.1 | - | 6 | 597 | |||
AK167834.1 | - | 6 | 601 | |||
AK205222.1 | - | 2 | 155 | |||
BC043133.1 | - | 6 | 524 | |||
BC063105.1 | - | 7 | 766 | 5003026 | ||
BC065171.1 | - | 6 | 553 | 6415034 | ||
BC090624.1 | - | 4 | 429 | 6815664 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001011988, XM_006241369, XM_006241366, XM_006241365, XM_006241367, and others.
Gene Ontology Categories:
