siRNA Details
Gene Symbol: Ints12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027927.3 | NP_082203.1 | 4 | 377 | ||
GenBank mRNA | AK003874.1 | - | 4 | 389 | ||
AK012829.1 | - | 4 | 390 | |||
AK038713.1 | - | 2 | 3944 | |||
AK054389.1 | - | 1 | 1250 | |||
AK155162.1 | - | 4 | 389 | |||
BC010657.1 | - | 2 | 49 | |||
BC023801.1 | - | 4 | 377 | 5353058 | ||
BC046996.1 | - | 4 | 371 | 6494551 |
