siRNA Details
Gene Symbol: Dpep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027960.2 | NP_082236.1 | 9 | 1240 | ||
GenBank mRNA | AF488553.1 | - | 9 | 1121 | ||
AK006085.1 | - | 9 | 1241 | |||
BC051148.1 | - | 9 | 1250 | 6704315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
