siRNA Details
Gene Symbol: Tmem106b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027992.3 | NP_082268.2 | 6 | 856 | ||
GenBank mRNA | AK009646.1 | - | 6 | 759 | ||
AK148477.1 | - | 6 | 780 | |||
BC050246.1 | - | 6 | 773 | 6313498 |
