siRNA Details
Gene Symbol: Snx10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127348.1 | NP_001120820.1 | 7 | 867 | ||
NM_001127349.1 | NP_001120821.1 | 7 | 872 | |||
NM_028035.4 | NP_082311.3 | 7 | 970 | |||
GenBank mRNA | AK010399.1 | - | 7 | 815 | ||
AK028163.1 | - | 7 | 971 | |||
AK150704.1 | - | 7 | 875 | |||
AK150878.1 | - | 7 | 789 | |||
AK150999.1 | - | 7 | 875 | |||
AK151320.1 | - | 7 | 875 | |||
AK151520.1 | - | 7 | 875 | |||
AK152004.1 | - | 7 | 793 | |||
AK152825.1 | - | 7 | 847 | |||
AK152949.1 | - | 7 | 863 | |||
AK153127.1 | - | 7 | 875 | |||
AK153356.1 | - | 7 | 825 | |||
AK166121.1 | - | 7 | 840 | |||
AK169110.1 | - | 7 | 806 | |||
BC010334.1 | - | 7 | 804 | 2655824 |
