siRNA Details
Gene Symbol: Tmco6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028036.3 | NP_082312.3 | 12 | 1800 | ||
GenBank mRNA | AK010487.1 | - | 12 | 1724 | ||
BC005637.1 | - | 12 | 1799 | 3156734 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
