siRNA Details
Gene Symbol: Cnot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037846.3 | NP_001032935.2 | 16 | 1995 | ||
NM_001037847.2 | NP_001032936.2 | 16 | 1925 | |||
NM_028082.2 | NP_082358.2 | 15 | 1872 | |||
GenBank mRNA | AK011231.1 | - | 14 | 1795 | ||
AK078121.1 | - | 16 | 1985 | |||
AK149767.1 | - | 16 | 1969 | |||
AK167834.1 | - | 16 | 1973 | |||
BC043133.1 | - | 16 | 1896 | |||
BC063105.1 | - | 17 | 2138 | 5003026 | ||
BC065171.1 | - | 16 | 1925 | 6415034 | ||
BC090624.1 | - | 14 | 1801 | 6815664 |

More Information
Other Species Targeted

Gene Ontology Categories:
