siRNA Details
Gene Symbol: MTTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000253.3 | NP_000244.2 | 4 | 579 | ||
NM_001300785.1 | NP_001287714.1 | 3 | 417 | |||
GenBank mRNA | AK225328.1 | - | 3 | 404 | ||
AK290793.1 | - | 3 | 386 | |||
AK302602.1 | - | 3 | 414 | |||
BC062696.1 | - | 3 | 408 | |||
BC125110.1 | - | 3 | 358 | 40118807 | ||
BC125111.1 | - | 3 | 358 | 40118809 | ||
X59657.1 | - | 3 | 404 | |||
X75500.1 | - | 3 | 342 | |||
X91148.1 | - | 3 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
