siRNA Details
Gene Symbol: Zfp661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111029.1 | NP_001104499.1 | 3 | 214 | ||
NM_028141.3 | NP_082417.3 | 4 | 431 | |||
GenBank mRNA | AK033122.1 | - | 3 | 216 | ||
AK134651.1 | - | 4 | 1484 | |||
BC052876.1 | - | 4 | 435 | 30047253 |
