siRNA Details
Gene Symbol: Kalrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177357.3 | NP_796331.2 | 38, 39 | 5807 | ||
GenBank mRNA | AK088732.1 | - | 4, 5 | 754 | ||
AK139581.1 | - | 5, 6 | 1911 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_032062, AF232669, AF232668.
Gene Ontology Categories:
