siRNA Details
Gene Symbol: Pgm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028132.3 | NP_082408.3 | 3 | 615 | ||
GenBank mRNA | AK011485.1 | - | 3 | 540 | ||
AK147982.1 | - | 3 | 587 | |||
AK152678.1 | - | 3 | 615 | |||
AK152923.1 | - | 3 | 615 | |||
AK181726.1 | - | 2 | 274 | |||
AK199001.1 | - | 3 | 311 | |||
AK200679.1 | - | 2 | 242 | |||
AK203334.1 | - | 2 | 210 | |||
AK212160.1 | - | 2 | 230 | |||
AK212644.1 | - | 3 | 312 | |||
BC036290.1 | - | 3 | 803 | |||
BC055713.1 | - | 3 | 566 | |||
BC080801.1 | - | 3 | 586 | |||
BC086490.1 | - | 3 | 520 | 30435602 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
