siRNA Details
Gene Symbol: NF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000268.3 | NP_000259.1 | 3, 4 | 814 | ||
NM_016418.5 | NP_057502.2 | 3, 4 | 814 | |||
NM_181825.2 | NP_861546.1 | 3, 4 | 814 | |||
NM_181828.2 | NP_861966.1 | 2, 3 | 688 | |||
NM_181832.2 | NP_861970.1 | 3, 4 | 814 | |||
NM_181833.2 | NP_861971.1 | 3, 4 | 814 | |||
XM_017028809.1 | XP_016884298.1 | 3, 4 | 377 | |||
XM_017028810.1 | XP_016884299.1 | 3, 4 | 373 | |||
GenBank mRNA | AF122827.1 | - | 3, 4 | 421 | ||
AF123570.1 | - | 3, 4 | 421 | |||
AF369657.1 | - | 3, 4 | 796 | |||
AF369658.1 | - | 3, 4 | 796 | |||
AF369661.1 | - | 3, 4 | 371 | |||
AF369662.1 | - | 3, 4 | 371 | |||
AF369663.1 | - | 2, 3 | 245 | |||
AF369666.1 | - | 4, 5 | 487 | |||
AF369667.1 | - | 3, 4 | 396 | |||
AF369668.1 | - | 3, 4 | 371 | |||
AF369701.1 | - | 3, 4 | 737 | |||
AK297116.1 | - | 3, 4 | 753 | |||
BC020257.1 | - | 3, 4 | 709 | 4871980 | ||
CR456530.1 | - | 3, 4 | 433 | |||
L11353.1 | - | 3, 4 | 590 | |||
Z22664.1 | - | 3, 4 | 545 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
