siRNA Details
Gene Symbol: Kalrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177357.3 | NP_796331.2 | 38, 39 | 5811 | ||
GenBank mRNA | AK088732.1 | - | 4, 5 | 758 | ||
AK139581.1 | - | 5, 6 | 1915 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
