siRNA Details
Gene Symbol: NPHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000272.3 | NP_000263.2 | 5 | 534 | ||
NM_001128178.1 | NP_001121650.1 | 5 | 534 | |||
NM_001128179.1 | NP_001121651.1 | 3 | 348 | |||
NM_207181.2 | NP_997064.2 | 5 | 534 | |||
XM_005263676.1 | XP_005263733.1 | 5 | 483 | |||
XM_005263677.1 | XP_005263734.1 | 5 | 483 | |||
XM_005263678.2 | XP_005263735.1 | 5 | 483 | |||
XM_005263679.1 | XP_005263736.1 | 5 | 483 | |||
XM_006712551.1 | XP_006712614.1 | 5 | 483 | |||
XM_006712552.2 | XP_006712615.1 | 5 | 483 | |||
XM_011511244.1 | XP_011509546.1 | 5 | 483 | |||
XM_017004218.1 | XP_016859707.1 | 5 | 483 | |||
GenBank mRNA | AF023674.1 | - | 5 | 435 | ||
AJ001815.1 | - | 5 | 473 | |||
AK225600.1 | - | 5 | 464 | |||
AK299011.1 | - | 5 | 463 | |||
AK307654.1 | - | 5 | 467 | |||
BC062574.1 | - | 3 | 258 | 5187280 | ||
BX571745.1 | - | 5 | 490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
