siRNA Details
Gene Symbol: Strada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252448.1 | NP_001239377.1 | 13 | 1637 | ||
NM_001252449.1 | NP_001239378.1 | 10 | 1490 | |||
NM_028126.3 | NP_082402.1 | 11 | 1526 | |||
GenBank mRNA | AK011451.1 | - | 11 | 1472 | ||
AK138358.1 | - | 13 | 1638 | |||
AY341872.1 | - | 11 | 1480 | |||
AY341878.1 | - | 10 | 1444 | |||
BC052063.1 | - | 1533 | ||||
BC058517.1 | - | 11 | 1427 | 5717255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
