siRNA Details
Gene Symbol: Kalrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177357.3 | NP_796331.2 | 42, 43 | 6145 | ||
GenBank mRNA | AK088732.1 | - | 8, 9 | 1092 | ||
AK139581.1 | - | 9, 10 | 2249 |

More Information
Other Species Targeted

Gene Ontology Categories:
