siRNA Details
Gene Symbol: Zfp655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028298.3 | NP_082574.1 | 4 | 929 | ||
GenBank mRNA | AK012336.1 | - | 4 | 932 | ||
AK220552.1 | - | 4 | 934 | |||
BC019153.1 | - | 5 | 1025 | |||
BC034731.1 | - | 4 | 932 | |||
BC055296.1 | - | 1 | 331 | |||
BC060660.1 | - | 4 | 965 | 30355779 | ||
BC062945.1 | - | 4 | 929 | 30357186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
