siRNA Details
Gene Symbol: 3110082I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028469.3 | NP_082745.2 | 3 | 237 | ||
GenBank mRNA | AK014271.1 | - | 3 | 218 | ||
AV470085.1 | - | 3 | 191 | |||
BC044867.1 | - | 3 | 217 | |||
BC058580.1 | - | 3 | 230 | |||
BU562762.1 | - | 3 | 239 |
