siRNA Details
Gene Symbol: 1700040L02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028491.1 | NP_082767.1 | 1, 2 | 137 | ||
GenBank mRNA | AK006661.1 | - | 1, 2 | 138 | ||
AK029622.1 | - | 1, 2 | 137 | |||
BC087900.1 | - | 1, 2 | 127 | 6774064 |

More Information
Other Species Targeted

Gene Ontology Categories:
