siRNA Details
Gene Symbol: Aldh3b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028545.2 | NP_082821.2 | 10 | 1669 | ||
GenBank mRNA | AK006803.1 | - | 3 | 567 | ||
AK081788.1 | - | 9 | 2019 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
