siRNA Details
Gene Symbol: PLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000301.3 | NP_000292.1 | 19 | 2513 | ||
GenBank mRNA | AK298338.1 | - | 15 | 1869 | ||
AK312966.1 | - | 19 | 2464 | |||
BC060513.1 | - | 19 | 2439 | 4609795 | ||
BU073846.1 | - | 3 | 321 | |||
CR749293.1 | - | 6 | 4359 | |||
K02922.1 | - | 12 | 1528 | |||
M74220.1 | - | 19 | 2450 | |||
X05199.1 | - | 19 | 2455 |
Gene Symbol: EVA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF177337.1 | - | 68 |
Gene Symbol: LPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005577.2 | NP_005568.2 | 40 | 6211 | ||
GenBank mRNA | X06290.1 | - | 13660 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
