siRNA Details
Gene Symbol: Nphp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028721.3 | NP_082997.3 | 17, 18 | 2692 | ||
GenBank mRNA | AK014393.1 | - | 2, 3 | 252 | ||
AK133217.1 | - | 17, 18 | 2693 | |||
AK173327.1 | - | 17, 18 | 2271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
