siRNA Details
Gene Symbol: 3110082I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028469.3 | NP_082745.2 | 3 | 242 | ||
GenBank mRNA | AK014271.1 | - | 3 | 223 | ||
AV470085.1 | - | 3 | 196 | |||
BC044867.1 | - | 3 | 222 | |||
BC058580.1 | - | 3 | 235 | |||
BU562762.1 | - | 3 | 244 |
