siRNA Details
Gene Symbol: Aldh3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177438.1 | NP_001170909.1 | 10 | 1984 | ||
GenBank mRNA | AK048310.1 | - | 845 | |||
AK136505.1 | - | 10 | 1988 | |||
BC100370.1 | - | 842 |
Gene Symbol: Aldh3b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028545.2 | NP_082821.2 | 10 | 1818 | ||
GenBank mRNA | AK006803.1 | - | 3 | 716 | ||
AK081788.1 | - | 9 | 2168 |

More Information
Other Species Targeted

Gene Ontology Categories:
