siRNA Details
Gene Symbol: Marveld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028584.3 | NP_082860.2 | 3 | 949 | ||
GenBank mRNA | AK007346.1 | - | 3 | 870 | ||
BQ900249.1 | - | 3 | 590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
