siRNA Details
Gene Symbol: Dppa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018002.1 | NP_001018002.1 | 3 | 250 | ||
NM_028610.2 | NP_082886.2 | 3 | 250 | |||
GenBank mRNA | AF490348.1 | - | 3 | 167 | ||
AK010753.1 | - | 3 | 218 | |||
AK033197.1 | - | 3 | 251 | |||
BY731814.1 | - | 3 | 216 |
