siRNA Details
Gene Symbol: 1700040L02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028491.1 | NP_082767.1 | 6 | 672 | ||
GenBank mRNA | AK006661.1 | - | 6 | 673 | ||
AK029622.1 | - | 6 | 672 | |||
BC087900.1 | - | 6 | 662 | 6774064 |
