siRNA Details
Gene Symbol: Speer4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028561.2 | NP_082837.1 | 5 | 1887 | ||
GenBank mRNA | AK006975.1 | - | 5 | 1936 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
