siRNA Details
Gene Symbol: ATXN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000333.3 | NP_000324.1 | 3, 4 | 882 | ||
NM_001177387.1 | NP_001170858.1 | 2, 3 | 439 | |||
GenBank mRNA | AF032103.1 | - | 1, 2 | 21 | ||
AF032105.1 | - | 3, 4 | 882 | |||
AJ000517.1 | - | 3, 4 | 890 |
