siRNA Details
Gene Symbol: Acer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290543.1 | NP_001277472.1 | 6 | 1025 | ||
GenBank mRNA | AK010770.1 | - | 2 | 385 | ||
AK046888.1 | - | 462 | ||||
AK085306.1 | - | 6 | 1021 | |||
BC051923.1 | - | 422 | ||||
BC060063.1 | - | 422 | ||||
BC061463.1 | - | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
