siRNA Details
Gene Symbol: Dppa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018002.1 | NP_001018002.1 | 6 | 964 | ||
NM_028610.2 | NP_082886.2 | 7 | 1120 | |||
GenBank mRNA | AF490348.1 | - | 7 | 1037 | ||
AK010753.1 | - | 7 | 1088 | |||
AK033197.1 | - | 7 | 1121 | |||
BC064754.1 | - | 7 | 1059 | 30024281 |
