siRNA Details
Gene Symbol: Arpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177615.1 | NP_001171086.1 | 4, 5 | 632 | ||
NM_001177620.1 | NP_001171091.1 | 6, 7 | 579 | |||
NM_028755.3 | NP_083031.1 | 5, 6 | 541 | |||
GenBank mRNA | AK002950.2 | - | 5, 6 | 525 | ||
AK159004.1 | - | 4, 5 | 611 | |||
AK160860.1 | - | 6, 7 | 577 | |||
BC027107.1 | - | 5, 6 | 529 | |||
BY290692.1 | - | 4, 5 | 338 |

More Information
Other Species Targeted

Gene Ontology Categories:
