siRNA Details
Gene Symbol: Dock8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028785.3 | NP_083061.2 | 38 | 4984 | ||
GenBank mRNA | AK004816.2 | - | 2 | 122 | ||
AK028968.1 | - | 1 | 42 | |||
AK131180.1 | - | 15 | 1947 | |||
BC030316.1 | - | 15 | 1975 | |||
BC043470.1 | - | 7 | 787 | |||
BC055295.1 | - | 7 | 815 | |||
BC141358.1 | - | 38 | 4979 | 9055970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
