siRNA Details
Gene Symbol: Sipa1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081028.1 | NP_001074497.1 | 15, 16 | 4387 | ||
GenBank mRNA | AK044193.1 | - | 2, 3 | 214 | ||
AK087819.1 | - | 2, 3 | 372 | |||
AK163313.1 | - | 12, 13 | 2769 | |||
AK172968.1 | - | 8, 9 | 1130 | |||
BC095969.1 | - | 5, 6 | 920 | |||
BC144792.1 | - | 2, 3 | 293 | |||
BC145739.1 | - | 2, 3 | 293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
