siRNA Details
Gene Symbol: Mei1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028897.3 | NP_083173.2 | 11 | 1228 | ||
GenBank mRNA | AK016514.1 | - | 5 | 433 | ||
AY270177.1 | - | 4 | 974 | |||
BC137749.1 | - | 4 | 261 | 8860768 | ||
BC144888.1 | - | 4 | 261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
