siRNA Details
Gene Symbol: Cbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028763.3 | NP_083039.2 | 3, 4 | 310 | ||
GenBank mRNA | AK004679.2 | - | 3, 4 | 215 | ||
AK035250.1 | - | 3, 4 | 217 | |||
AK090253.1 | - | 3, 4 | 215 | |||
AK146048.1 | - | 3, 4 | 215 | |||
AK146103.1 | - | 3, 4 | 215 | |||
BC048240.1 | - | 3, 4 | 207 | 6493113 | ||
BY237361.1 | - | 3, 4 | 311 |
Gene Symbol: Npcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013360.2 | NP_001013378.2 | 3, 4 | 310 | ||
NM_001013362.2 | NP_001013380.2 | 3, 4 | 310 | |||
GenBank mRNA | AY394488.1 | - | 3, 4 | 215 | ||
AY394489.1 | - | 3, 4 | 215 | |||
AY395907.1 | - | 3, 4 | 215 |

More Information
Other Species Targeted

Gene Ontology Categories:
