siRNA Details
Gene Symbol: TGM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000359.2 | NP_000350.1 | 2, 3 | 453 | ||
GenBank mRNA | AK291350.1 | - | 2, 3 | 420 | ||
AK315819.1 | - | 2, 3 | 453 | |||
AK315828.1 | - | 2, 3 | 413 | |||
AK315843.1 | - | 2, 3 | 420 | |||
BC034699.1 | - | 2, 3 | 404 | 4747045 | ||
D90287.1 | - | 2, 3 | 409 | |||
DA437895.1 | - | 2, 3 | 453 | |||
M55183.1 | - | 2, 3 | 382 | |||
M62925.1 | - | 2, 3 | 415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
