siRNA Details
Gene Symbol: Dock8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028785.3 | NP_083061.2 | 41 | 5360 | ||
GenBank mRNA | AK004816.2 | - | 5 | 498 | ||
AK028968.1 | - | 4 | 418 | |||
AK131180.1 | - | 18 | 2323 | |||
BC030316.1 | - | 18 | 2351 | |||
BC043470.1 | - | 10 | 1163 | |||
BC055295.1 | - | 10 | 1191 | |||
BC141358.1 | - | 41 | 5355 | 9055970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
