siRNA Details
Gene Symbol: Sipa1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081028.1 | NP_001074497.1 | 16 | 4515 | ||
GenBank mRNA | AK044193.1 | - | 3 | 342 | ||
AK087819.1 | - | 3 | 500 | |||
AK163313.1 | - | 13 | 2897 | |||
AK172968.1 | - | 9 | 1258 | |||
BC095969.1 | - | 6 | 1048 | |||
BC116408.1 | - | 1 | 108 | |||
BC128493.1 | - | 1 | 108 | |||
BC144792.1 | - | 3 | 421 | |||
BC145739.1 | - | 3 | 421 | |||
BC152360.1 | - | 1 | 108 | |||
BC152361.1 | - | 1 | 96 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
