siRNA Details
Gene Symbol: TPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000366.5 | NP_000357.3 | 2 | 352 | ||
NM_001018004.1 | NP_001018004.1 | 2 | 352 | |||
NM_001018005.1 | NP_001018005.1 | 2 | 352 | |||
NM_001018006.1 | NP_001018006.1 | 2 | 352 | |||
XM_005254637.1 | XP_005254694.1 | 3 | 433 | |||
XM_005254638.3 | XP_005254695.2 | 2 | 795 | |||
XM_005254639.3 | XP_005254696.2 | 2 | 795 | |||
XM_005254645.2 | XP_005254702.2 | 2 | 795 | |||
XM_006720667.3 | XP_006720730.2 | 2 | 795 | |||
XM_017022534.1 | XP_016878023.1 | 2 | 795 | |||
XM_017022536.1 | XP_016878025.1 | 2 | 795 | |||
XM_017022538.1 | XP_016878027.1 | 2 | 795 | |||
XM_017022539.1 | XP_016878028.1 | 2 | 795 | |||
GenBank mRNA | AK129882.1 | - | 2 | 240 | ||
AK131384.1 | - | 3 | 366 | |||
AK299387.1 | - | 2 | 241 | |||
AK309170.1 | - | 2 | 561 | |||
AY640415.1 | - | 2 | 161 | |||
BC007433.2 | - | 2 | 302 | 2962925 | ||
BG107949.1 | - | 2 | 359 | |||
CN423344.1 | - | 2 | 290 | |||
GQ472198.1 | - | 2 | 302 | |||
KU178366.1 | - | 2 | 161 | |||
KU178367.1 | - | 2 | 161 | |||
M19713.1 | - | 2 | 217 | |||
M19714.1 | - | 2 | 103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
