siRNA Details
Gene Symbol: Nsun6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165941.1 | NP_001159413.1 | 5 | 770 | ||
NM_001165943.1 | NP_001159415.1 | 4 | 474 | |||
NM_028950.4 | NP_083226.3 | 4 | 657 | |||
GenBank mRNA | AK016629.1 | - | 4 | 475 | ||
AK016811.1 | - | 5 | 783 | |||
AK077778.1 | - | 4 | 541 | |||
AK132428.1 | - | 4 | 624 | |||
BC053751.1 | - | 4 | 625 | 6335896 |
