siRNA Details
Gene Symbol: Taf7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028958.4 | NP_083234.2 | 5 | 674 | ||
GenBank mRNA | AF285574.1 | - | 5 | 478 | ||
AK017109.1 | - | 5 | 675 | |||
BC106854.2 | - | 5 | 466 | 40046501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
