siRNA Details
Gene Symbol: Dock8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028785.3 | NP_083061.2 | 48 | 7177 | ||
GenBank mRNA | AK004816.2 | - | 12 | 2315 | ||
AK028968.1 | - | 11 | 2235 | |||
BC029018.1 | - | 7 | 1586 | |||
BC043470.1 | - | 17 | 2980 | |||
BC055295.1 | - | 17 | 3008 | |||
BC141358.1 | - | 48 | 7172 | 9055970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
