siRNA Details
Gene Symbol: 8430419L09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028982.4 | NP_083258.2 | 3, 4 | 618 | ||
GenBank mRNA | AK018428.1 | - | 3, 4 | 562 | ||
AK039401.1 | - | 3, 4 | 554 | |||
AK199895.1 | - | 1, 2 | 49 | |||
AK220395.1 | - | 2, 3 | 268 | |||
BC038002.1 | - | 2, 3 | 279 | |||
BC079676.1 | - | 3, 4 | 545 | |||
BC137674.1 | - | 3, 4 | 597 | 8860693 | ||
BC137676.1 | - | 3, 4 | 597 | 8860695 |
