siRNA Details
Gene Symbol: Iqca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029122.2 | NP_083398.2 | 2 | 339 | ||
GenBank mRNA | AK015512.1 | - | 2 | 340 | ||
AK016353.1 | - | 2 | 340 |

More Information
Other Species Targeted

Gene Ontology Categories:
