siRNA Details
Gene Symbol: Spag16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271533.1 | NP_001258462.1 | 1, 2 | 162 | ||
NM_029160.3 | NP_083436.2 | 1, 2 | 162 | |||
GenBank mRNA | AF490391.1 | - | 1, 2 | 144 | ||
AK015884.1 | - | 1, 2 | 163 | |||
AK083083.1 | - | 1, 2 | 141 | |||
BC120668.1 | - | 1, 2 | 161 | 40129591 | ||
BC145006.1 | - | 1, 2 | 161 | 9053554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
