siRNA Details
Gene Symbol: Arhgap24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029270.2 | NP_083546.2 | 2 | 582 | ||
GenBank mRNA | AK031879.1 | - | 2 | 169 | ||
AK052395.1 | - | 2 | 445 | |||
AK080986.1 | - | 2 | 582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
